Anti-PTEN Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101096
89318-354EA 582.76 USD
89318-354
Anti-PTEN Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to cGK1 (protein kinase, cGMP-dependent, type I)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PTEN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR