Anti-UCHL1 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX101093
89318-350EA 427.1 USD
89318-350
Anti-UCHL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PGP9.5 (ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase))

Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR