Anti-FGFR4 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX101054
89318-346EA 394.91 USD
89318-346
Anti-FGFR4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to alpha 1 Antitrypsin (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SERPINA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR