Anti-PIRH2 Rabbit Polyclonal Antibody

GTX101049
89330-376EA 376.43 USD
89330-376
Anti-PIRH2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SHC1 (SHC (Src homology 2 domain containing) transforming protein 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PIRH2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR