Anti-ICAM1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10103
89287-836EA 607.87 USD
89287-836
Anti-ICAM1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody to SH3BP1

Approx 70kDa band observed in Human Lung and Kidney lysates (predicted MW of 71kDa according to NP_061830). Recommended for use at 1-3µg/ml. In paraffin embedded Human Kidney, stains the cytoplasm of lymphoid cells in the interstitial parenchyma and of lymphocytes in a glomerulus. Recommended concentration, 0.3µg/ml.

Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR