Anti-NPT Rabbit Polyclonal Antibody

GTX100900
89316-366EA 376.43 USD
89316-366
Anti-NPT Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to CRK (v-crk sarcoma virus CT10 oncogene homolog (avian))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: NPT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR