Anti-TIMP3 Mouse Monoclonal Antibody [clone: 43-2]

Supplier: GeneTex
GTX10090
89336-476EA 759 USD
89336-476
Anti-TIMP3 Mouse Monoclonal Antibody [clone: 43-2]
Antibodies
Mouse monoclonal [43 - 2] to IL2

ELISA: Use at an assay dependent dilution. Suitable for sandwich ELISA of IL2 in the following combination: capture: GTX10090 / conjugate: GTX10089. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TIMP3
Clonality: Monoclonal
Clone: 43-2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR