Anti-GLP Mouse Monoclonal Antibody [clone: 4E6]

Supplier: GeneTex
GTX10084
89336-474EA 759 USD
89336-474
Anti-GLP Mouse Monoclonal Antibody [clone: 4E6]
Antibodies
Mouse monoclonal [4E6] to IL3

ELISA: Use at an assay dependent dilution. Suitable for sandwich ELISA of IL3 in the following combination: capture - GTX10084, 10080, 10083, conjugate - 10082. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IL3
Clonality: Monoclonal
Clone: 4E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR