Anti-KSHV ORF45 Mouse Monoclonal Antibody [clone: 2F2]

Supplier: GeneTex
GTX10080
89336-470EA 759 USD
89336-470
Anti-KSHV ORF45 Mouse Monoclonal Antibody [clone: 2F2]
Antibodies
Mouse monoclonal [2F2] to IL3

ELISA: Use at an assay dependent dilution. Suitable for sandwich ELISA of IL3 in the following combination: capture - GTX10084, 10080, 10083, conjugate - 10082. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: KSHV ORF45
Clonality: Monoclonal
Clone: 2F2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR