Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CHD3 Goat Polyclonal Antibody

Supplier: Genetex
GTX10079
89284-990
Anti-CHD3 Goat Polyclonal Antibody
Goat polyclonal antibody to Syntrophin gamma 1

Preliminary experiments gave no signal but low background in Human Testis and Brain extracts at up to 1µg/ml. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates?

Type: Primary
Antigen: CHD3
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty