Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-4E-BP1 Mouse Monoclonal Antibody [clone: GD4]

Supplier: Genetex

GTX10076
89336-468EA 246.49 USD InStock
89336-468
Anti-4E-BP1 Mouse Monoclonal Antibody [clone: GD4]
Antibodies
Mouse monoclonal antibody [GD4] to IFN gamma

ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 20 kDa. GTX28318 is recommended to be used for coating and GTX10075 and GTX10076 are recommended to be used as conjugates. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: 4E-BP1
Clonality: Monoclonal
Clone: GD4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty