Anti-4E-BP1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100759
89318-304EA 597.31 USD
89318-304
Anti-4E-BP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Retinoic Acid Receptor beta (retinoic acid receptor, beta)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: 4E-BP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR