Anti-RSPO1 Mouse Monoclonal Antibody [clone: 4A3]

Supplier: GeneTex
GTX10071
89336-798EA 659.34 USD
89336-798
Anti-RSPO1 Mouse Monoclonal Antibody [clone: 4A3]
Antibodies
Mouse monoclonal [4A3] to alpha Fetoprotein

ELISA: Use at an assay dependent dilution. Can be used in an ELISA pair (GTX10071 as capture antibody and GTX10072 as detection antibody). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: RSPO1
Clonality: Monoclonal
Clone: 4A3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR