Anti-SQSTM1 / P62 Antibody [N3C1]

Supplier: GeneTex
GTX100685
89318-294EA 597.31 USD
89318-294
Anti-SQSTM1 / P62 Antibody [N3C1]
Antibodies
Rabbit Polyclonal antibody to SQSTM1 (sequestosome 1)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, IP: 1:100-1:500, FACS. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SQSTM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR