Anti-ALCAM Rabbit Polyclonal Antibody

Supplier: Genetex

GTX100654
89318-288EA 395.32 USD
89318-288
Anti-ALCAM Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MDM2 (Mdm2 p53 binding protein homolog)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ALCAM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR