Anti-HDAC3 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX100600
89307-822EA 793.49 USD
89307-822
Anti-HDAC3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PTP4A3 (protein tyrosine phosphatase type IVA, member 3)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: HDAC3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR