Anti-HDAC3 Mouse Monoclonal Antibody [clone: F23]

Supplier: GeneTex
GTX10060
89336-460EA 759 USD
89336-460
Anti-HDAC3 Mouse Monoclonal Antibody [clone: F23]
Antibodies
Mouse monoclonal [F23] to Ferritin

ELISA: Use at an assay dependent dilution. Can be used with GTX10061 (conjugate). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HDAC3
Clonality: Monoclonal
Clone: F23
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR