Anti-LIN28 Rabbit Polyclonal Antibody

GTX100576
89307-808EA 176.03 USD
89307-808
Anti-LIN28 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to ICAM5 (intercellular adhesion molecule 5, telencephalin)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: LIN28
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR