Anti-NCOA3 Mouse Monoclonal Antibody [clone: DD3]

Supplier: GeneTex
GTX10053
89336-450EA 759 USD
89336-450
Anti-NCOA3 Mouse Monoclonal Antibody [clone: DD3]
Antibodies
Mouse monoclonal [DD3] to D - Dimer

ELISA: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Can be paired for ELISA with GTX10055 Mouse monoclonal [DD5] to D-Dimer . To be analyzed in sandwich immunoassay, plasma must be diluted with 10 mM Tris/HCl buffer, pH 7.5, containing 1 M NaCl and 0.1 % Tween 20. To avoid non-specific binding, the final NaCl concentration in plasma samples must be 0.5 M or more. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: NCOA3
Clonality: Monoclonal
Clone: DD3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR