Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-F8 Mouse Monoclonal Antibody [clone: DD2]

Supplier: Genetex

GTX10052
89336-448EA 246.49 USD InStock
89336-448
Anti-F8 Mouse Monoclonal Antibody [clone: DD2]
Antibodies
Mouse monoclonal [DD2] to D - Dimer

ELISA: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 0 kDa. Can be paired for ELISA with GTX10054 Mouse monoclonal [DD4] to D-Dimer , GTX10055 Mouse monoclonal [DD5] to D-Dimer and GTX10056 Mouse monoclonal [DD6] to D-Dimer . To be analyzed in sandwich immunoassay, plasma must be diluted with 10 mM Tris/HCl buffer, pH 7.5, containing 1 M NaCl and 0.1 % Tween 20. To avoid non-specific binding, the final NaCl concentration in plasma samples must be 0.5 M or more. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: DD2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty