Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DKK1 Mouse Monoclonal Antibody [clone: DD1]

Supplier: Genetex
GTX10050
89336-444
Anti-DKK1 Mouse Monoclonal Antibody [clone: DD1]
Mouse monoclonal [DD1] to D - Dimer

ELISA: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Can be paired for ELISA with this antibody as capture and GTX10054 Mouse monoclonal [DD4] to D-Dimer , GTX10055 Mouse monoclonal [DD5] to D-Dimer or GTX10056 Mouse monoclonal [DD6] to D-Dimer as detection. Less than 10ng/ml of D-dimer can be detected using this antibody and HRP labeled GTX10056 [DD6]. To be analyzed in sandwich immunoassay, plasma must be diluted with 10 mM Tris/HCl buffer, pH 7.5, containing 1 M NaCl and 0.1 % Tween 20. To avoid non-specific binding, the final NaCl concentration in plasma samples must be 0.5 M or more. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: DKK1
Clonality: Monoclonal
Clone: DD1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty