Anti-KLF15 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX100453
89307-770EA 823.81 USD
89307-770
Anti-KLF15 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to IGF2 (insulin-like growth factor 2 (somatomedin A))

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: KLF15
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR