Anti-ESR1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX10043
89277-278EA 355.14 USD
89277-278
Anti-ESR1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to VPS33A

Peptide ELISA: antibody detection limit dilution 1/16000. Western Blot: Preliminary experiments gave no signal but low background in Human Brain and Molt-4 extracts at up to 1?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ESR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR