Anti-EPHA5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100366
89318-232EA 582.76 USD
89318-232
Anti-EPHA5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GPR20 (G protein-coupled receptor 20)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: EPHA5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR