Anti-IGF1R Rabbit Polyclonal Antibody

Supplier: Genetex

GTX100300
89318-204EA 394.91 USD
89318-204
Anti-IGF1R Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Glycophorin A (glycophorin A (MNS blood group))

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: IGF1R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR