Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BNP Mouse Monoclonal Antibody [clone: C4]

Supplier: Genetex
GTX10028
89336-438
Anti-BNP Mouse Monoclonal Antibody [clone: C4]
Mouse monoclonal [C4] to C Reactive Protein

AP: Use at an assay dependent dilution. ELISA: Use at an assay dependent dilution. IA: Use at an assay dependent dilution. Can be paired for ELISA with GTX28277 Mouse monoclonal [C2] to C Reactive Protein . Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: BNP
Clonality: Monoclonal
Clone: C4
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty