Anti-CTTN Rabbit Polyclonal Antibody

Supplier: Genetex

GTX100253
89318-192EA 394.91 USD
89318-192
Anti-CTTN Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Cortactin (src substrate cortactin)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CTTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR