Anti-PTPRC Rabbit Polyclonal Antibody

Supplier: Genetex

GTX100052
89318-148EA 394.91 USD
89318-148
Anti-PTPRC Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Survivin (baculoviral IAP repeat containing 5)

Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: PTPRC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR