Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Human;Mouse;Rat;Monkey AKT Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemical

200-306-269
RL200306269EA 321.26 USD InStock
RL200306269
Goat Anti-Human;Mouse;Rat;Monkey AKT Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: AKT
Clonality: Polyclonal
Clone: 18F3.H11
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat, Monkey

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty