Anti-AKT2 Rat Monoclonal Antibody [clone: 16G11.A7]

Supplier: Rockland Immunochemicals
200-301-096S
RL200301096SEA 147.5 USD
RL200301096S
Anti-AKT2 Rat Monoclonal Antibody [clone: 16G11.A7]
Antibodies
Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 16G11.A7
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR