Anti-CD11a Rat Monoclonal Antibody [clone: M17/4]

Supplier: BioLegend
101118
101118-BLEA 160.97 USD
101118-BL
Anti-CD11a Rat Monoclonal Antibody [clone: M17/4]
Antibodies
Ultra-LEAF™ Purified anti-mouse CD11a [M17/4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, Block, IHC, IP; Size: 1 mg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume or 100 μl of whole blood.  It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone: M17/4
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR