Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

Anti-ST3 beta-galactoside alpha-2,3-sialyltransferase 3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP9999B
89399-540EA 401.06 USD InStock
89399-540
Anti-ST3 beta-galactoside alpha-2,3-sialyltransferase 3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ST3 beta-galactoside alpha-2,3-sialyltransferase 3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty