Anti-PPP1R14B Rabbit Polyclonal Antibody

Supplier: Abgent
AP9988A
89399-518EA 436.86 USD
89399-518
Anti-PPP1R14B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PPP1R14B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR