Anti-C19orf33 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9983A
89399-508EA 436.86 USD
89399-508
Anti-C19orf33 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: C19orf33
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR