Anti-MRAP Rabbit Polyclonal Antibody

Supplier: Abgent
AP9980A
89399-502EA 436.86 USD
89399-502
Anti-MRAP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MRAP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR