Anti-GALNT5 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9976A
89399-494EA 610.34 USD
89399-494
Anti-GALNT5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GALNT5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR