Anti-DPAGT1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP9967A
89399-476EA 436.86 USD
89399-476
Anti-DPAGT1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DPAGT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR