Anti-CENPM Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9955A
89399-452EA 580.97 USD
89399-452
Anti-CENPM Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CENPM
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR