Anti-HCCS Rabbit Polyclonal Antibody

Supplier: Abgent
AP9883C
89399-304EA 436.73 USD
89399-304
Anti-HCCS Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HCCS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR