Anti-BSND Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9858B
89399-256EA 610.01 USD
89399-256
Anti-BSND Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BSND
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR