Anti-CK78 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9856C
89399-252EA 610.01 USD
89399-252
Anti-CK78 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CK78
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR