Anti-CCDC28B Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9855A
89399-250EA 436.86 USD
89399-250
Anti-CCDC28B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCDC28B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR