Anti-GLG1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9839B
89399-218EA 436.73 USD
89399-218
Anti-GLG1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GLG1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR