Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ST8 Alpha-N-Acetyl-Neuraminide Alpha-2,8-Sialyltransferase 4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP9771C
89399-082EA 401.06 USD InStock
89399-082
Anti-ST8 Alpha-N-Acetyl-Neuraminide Alpha-2,8-Sialyltransferase 4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ST8 Alpha-N-Acetyl-Neuraminide Alpha-2,8-Sialyltransferase 4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty