Anti-GAGE12F Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9756B
89399-050EA 436.86 USD
89399-050
Anti-GAGE12F Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GAGE12F; GAGE7; GAGE12I; GAGE12G
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR