Anti-TM4SF4 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9749A
89399-036EA 610.01 USD
89399-036
Anti-TM4SF4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TM4SF4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR