Anti-NAP1L5 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9744C
89399-026EA 436.86 USD
89399-026
Anti-NAP1L5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NAP1L5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR