Anti-CD7 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9733C
89399-004EA 610.01 USD
89399-004
Anti-CD7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CD7
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR