Anti-CLDND1 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9729C
89398-994EA 436.86 USD
89398-994
Anti-CLDND1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CLDND1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR