Anti-ADP-Ribosylhydrolase like 2 Rabbit Polyclonal Antibody

Supplier: Abcepta
AP9723A
89398-982EA 609.9 USD
89398-982
Anti-ADP-Ribosylhydrolase like 2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: [delete]
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR